2-Thenoesäure
Lieferant: Molekula
Synonyme:
Thiophen-2-carbonsäure, 2-Thiophencarbonsäure
26032569-100G
MOLE26032569-100GEA
0
EUR
MOLE26032569-100G
2-Thenoesäure
2-Thenoesäure
Formel:
C₅H₄O₂S Molecular Weight: 128,15 g/mol Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 123…125 °C |
MDL:
MFCD00005437 CAS-Nummer: 527-72-0 EINECS: 208-423-4 Merck Index: 12,09491 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...