Lansoprazol

Lieferant: Molekula

Synonyme: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

26059460-1G
MOLE26059460-1GEA 219 EUR
MOLE26059460-1G
Lansoprazol
Lansoprazol
Formel: C₁₆H₁₄F₃N₃O₂S
Molecular Weight: 369.36 g/mol
MDL: MFCD00866873
CAS-Nummer: 103577-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR