5-Nitrochinolin

Lieferant: Molekula

26220752-5G
MOLE26220752-5GEA 0 EUR
MOLE26220752-5G
5-Nitrochinolin
5-Nitroquinoline
Formel: C₉H₆N₂O₂
Molecular Weight: 174,16 g/mol
Siedepunkt: 323 °C (1013 hPa)
Schmelzpunkt: 71 °C
MDL: MFCD00006790
CAS-Nummer: 607-34-1
EINECS: 210-134-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR