3-Brom-4-fluorbenzoesäure

Lieferant: Molekula
Achtung

26314892-1G
MOLE26314892-1GEA 0 EUR
MOLE26314892-1G
3-Brom-4-fluorbenzoesäure
3-Bromo-4-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD00042463
CAS-Nummer: 1007-16-5
EINECS: 213-751-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR