2,4,6-Trimethoxybenzaldehyd
Lieferant: Molekula
26543568-25G
MOLE26543568-25GEA
0
EUR
MOLE26543568-25G
2,4,6-Trimethoxybenzaldehyd
2,4,6-Trimethoxybenzaldehyde
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol Siedepunkt: 291 °C (1013 hPa) Schmelzpunkt: 120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003313 CAS-Nummer: 830-79-5 EINECS: 212-598-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...