2-Fluor-α-methyl-4-biphenylessigsäure

Lieferant: Molekula

Synonyme: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

26685423-1G
MOLE26685423-1GEA 0 EUR
MOLE26685423-1G
2-Fluor-α-methyl-4-biphenylessigsäure
2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
MDL: MFCD00079303
CAS-Nummer: 5104-49-4
EINECS: 225-827-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR