Ammonium-8-anilinonaphthalin-1-sulfonat

Lieferant: Molekula

Synonyme: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4

26829370-100G
MOLE26829370-100GEA 0 EUR
MOLE26829370-100G
Ammonium-8-anilinonaphthalin-1-sulfonat
Ammonium-8-anilinonaphthalin-1-sulfonat
Formel: C₁₆H₁₆N₂O₃S
Molecular Weight: 316,37 g/mol
MDL: MFCD00012560
CAS-Nummer: 28836-03-5
EINECS: 249-265-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR