3,5-Dihydroxy-4-methylbenzoesäure

Lieferant: Molekula

27046613-5G
MOLE27046613-5GEA 0 EUR
MOLE27046613-5G
3,5-Dihydroxy-4-methylbenzoesäure
3,5-Dihydroxy-4-methylbenzoic acid
Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
MDL: MFCD00002514
CAS-Nummer: 28026-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR