(R)-(+)-4-Benzyl-2-oxazolidinon

Lieferant: Molekula

Synonyme: (R)-4-Benzyl-2-oxazolidinone

27204332-5G
MOLE27204332-5GEA 93.4 EUR
MOLE27204332-5G
(R)-(+)-4-Benzyl-2-oxazolidinon
(R)-(+)-4-Benzyl-2-oxazolidinon
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
MDL: MFCD00010846
CAS-Nummer: 102029-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR