(S)-(-)-Indolin-2-carbonsäure

Lieferant: Molekula

Synonyme: (S)-Indoline-2-carboxylic acid

27353427-5G
MOLE27353427-5GEA 0 EUR
MOLE27353427-5G
(S)-(-)-Indolin-2-carbonsäure
(S)-(-)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
MDL: MFCD00070578
CAS-Nummer: 79815-20-6
EINECS: 410-860-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR