3,3'-Diaminobenzidin Tetrahydrochlorid (DAB)
Lieferant: Molekula
Synonyme:
3,3',4,4'-Biphenyltetramine tetrahydrochloride, DAB HCl
27393749
MOLE27393749EA
0
EUR
MOLE27393749
3,3'-Diaminobenzidin Tetrahydrochlorid (DAB)
3,3'-Diaminobenzidin Tetrahydrochlorid
Formel:
C₁₂H₁₈Cl₄N₄ Molecular Weight: 360,11 g/mol |
MDL:
MFCD00012969 CAS-Nummer: 7411-49-6 EINECS: 231-018-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...