D(+)-α-Acetobromglucose
Lieferant: Molekula
Synonyme:
2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose
27442186-25G
MOLE27442186-25GEA
215
EUR
MOLE27442186-25G
D(+)-α-Acetobromglucose
D(+)-α-Acetobromglucose
Formel:
C₁₄H₁₉BrO₉ Molecular Weight: 411.2 g/mol |
MDL:
MFCD00063254 CAS-Nummer: 572-09-8 EINECS: 209-339-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...