5-Fluor-2-indolcarbonsäure

Lieferant: Molekula

Synonyme: 5-Fluoro-1H-indole-2-carboxylic acid

27753742-5G
MOLE27753742-5GEA 0 EUR
MOLE27753742-5G
5-Fluor-2-indolcarbonsäure
5-Fluoroindole-2-carboxylic acid
Formel: C₉H₆FNO₂
Schmelzpunkt: 258…259 °C
MDL: MFCD00005612
CAS-Nummer: 399-76-8
EINECS: 206-919-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR