3-Methylveratrol

Lieferant: Molekula
Achtung

Synonyme: 1,2-Dimethoxy-3-methylbenzol

27821470-25G
MOLE27821470-25GEA 0 EUR
MOLE27821470-25G
3-Methylveratrol
3-Methylveratrol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00008379
CAS-Nummer: 4463-33-6
EINECS: 224-726-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR