Isoscopoletin (6-hydroxy-4-methylcoumarin)

Lieferant: Molekula

Synonyme: 6-Hydroxy-4-methyl-2H-chromen-2-one, 6-Hydroxy-4-methylcoumarin

27832217-100MG
MOLE27832217-100MGEA 0 EUR
MOLE27832217-100MG
Isoscopoletin (6-hydroxy-4-methylcoumarin)
Isoscopoletin (6-Hydroxy-4-methylcoumarin)
Formel: C₁₀H₈O₃
Molecular Weight: 176,17 g/mol
MDL: MFCD00016967
CAS-Nummer: 2373-31-1
EINECS: 219-146-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR