5-Cyan-2-thiophenboronsäure

Lieferant: Molekula

Synonyme: (5-Cyanothiophen-2-yl)boronic acid, 5-Cyanothiophene-2-boronic acid

27885742-1G
MOLE27885742-1GEA 0 EUR
MOLE27885742-1G
5-Cyan-2-thiophenboronsäure
5-Cyano-2-thiopheneboronic acid
Formel: C₅H₄BNO₂S
Molecular Weight: 152,97 g/mol
MDL: MFCD02094029
CAS-Nummer: 305832-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR