5-Fluorisatin

Lieferant: Molekula
Achtung

Synonyme: 5-Fluor-1H-indol-2,3-dion, 5-Fluorindolin-2,3-dion, 5-Fluor-2,3-indolindion, 5-Fluor-2,3-dioxoindolin

27999310-5G
MOLE27999310-5GEA 71.6 EUR
MOLE27999310-5G
5-Fluorisatin
5-Fluorisatin
Formel: C₈H₄FNO₂
Molecular Weight: 165,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00022795
CAS-Nummer: 443-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR