(-)-Dimethyl-D-tartrat
Lieferant: Molekula
Synonyme:
(S,S)-(-)-2,3-Dihydroxybutandisäuredimethylester, D(-)-Threarsäuredimethylester, (S,S)-(-)-Weinsäuredimethylester, (S,S)-(-)-Dimethyltartrat, (-)-Dimethyl-D-tartrat, (S,S)-(-)-2,3-Dihydroxy-1,4-butandisäuredimethylester, (S,S)-(-)-2,3-Dihydroxybernsteinsäuredimethylester, D(-)-Weinsäuredimethylester
28005352-5G
MOLE28005352-5GEA
0
EUR
MOLE28005352-5G
(-)-Dimethyl-D-tartrat
(-)-Dimethyl-D-tartrat
Formel:
C₆H₁₀O₆ Molecular Weight: 178,14 g/mol Siedepunkt: 245 °C (1013 hPa) Schmelzpunkt: 45…48 °C |
MDL:
MFCD00008445 CAS-Nummer: 13171-64-7 EINECS: 236-118-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...