1-Fluorencarbonsäure

Lieferant: Molekula

28108738-1G
MOLE28108738-1GEA 0 EUR
MOLE28108738-1G
1-Fluorencarbonsäure
1-Fluorencarbonsäure
Formel: C₁₄H₁₀O₂
Molecular Weight: 210,23 g/mol
MDL: MFCD00001130
CAS-Nummer: 6276-03-5
EINECS: 228-472-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR