5-Nitrobenzimidazol

Lieferant: Molekula

Synonyme: 5-Nitro-1H-benzimidazole

28330984-100G
MOLE28330984-100GEA 0 EUR
MOLE28330984-100G
5-Nitrobenzimidazol
5-Nitrobenzimidazole
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
Schmelzpunkt: 200…203 °C
MDL: MFCD00005604
CAS-Nummer: 94-52-0
EINECS: 202-341-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR