1,2,3,4-Tetrahydrochinolin

Lieferant: Molekula

28354755-100G
MOLE28354755-100GEA 0 EUR
MOLE28354755-100G
1,2,3,4-Tetrahydrochinolin
1,2,3,4-Tetrahydrochinolin
Formel: C₉H₁₁N
Molecular Weight: 133,19 g/mol
MDL: MFCD00006693
CAS-Nummer: 635-46-1
EINECS: 211-237-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR