3-Propylphenol

Lieferant: Molekula
Gefahr

Synonyme: 3-N-Propylphenol

28380943-25G
MOLE28380943-25GEA 594 EUR
MOLE28380943-25G
3-Propylphenol
3-Propylphenol
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
MDL: MFCD01632130
CAS-Nummer: 621-27-2
EINECS: 210-675-5
UN: 3145
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR