5-Methylbenzotriazol

Lieferant: Molekula

Synonyme: 5-Methyl-1H-benzotriazole

28528039-100G
MOLE28528039-100GEA 0 EUR
MOLE28528039-100G
5-Methylbenzotriazol
5-Methylbenzotriazol
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Siedepunkt: 361 °C (1013 hPa)
Schmelzpunkt: 85 °C
MDL: MFCD00005702
CAS-Nummer: 136-85-6
EINECS: 205-265-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR