cis-4-Hydroxy-L(-)-prolin

Lieferant: Molekula

Synonyme: L(-)-cis-4-Hydroxyprolin, H-cis-Hyp-OH, (S)-(-)-cis-4-Hydroxypyrrolidin-2-carbonsäure, (S)-(-)-cis-4-Hydroxyprolin, (S)-(-)-cis-4-Hydroxyazolidin-2-carbonsäure, (S)-(-)-cis-4-Hydroxy-1,3,4,5-tetrahydro-2H-pyrrol- 2-carbonsäure, (2S)-(-)-cis-4-Hydroxypyrrolidin-2-carbonsäure

28587393-1G
MOLE28587393-1GEA 0 EUR
MOLE28587393-1G
cis-4-Hydroxy-L(-)-prolin
cis-4-Hydroxy-L(-)-prolin
Formel: C₅H₉NO₃
Molecular Weight: 131,13 g/mol
Schmelzpunkt: 257 °C
MDL: MFCD00064319
CAS-Nummer: 618-27-9
EINECS: 210-542-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR