PIPES Dinatriumsalz

Lieferant: Molekula

28635108-100G
MOLE28635108-100GEA 0 EUR
MOLE28635108-100G
PIPES Dinatriumsalz
PIPES Dinatriumsalz
Formel: C₈H₁₆N₂Na₂O₆S₂
MDL: MFCD00064349
CAS-Nummer: 76836-02-7
EINECS: 278-562-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR