Sudanorange G
Lieferant: Molekula
Synonyme:
4-(Phenylazo)resorcin, Solvent Orange 1
28640974-25G
MOLE28640974-25GEA
0
EUR
MOLE28640974-25G
Sudanorange G
Sudan Orange G
Formel:
C₁₂H₁₀N₂O₂ Molecular Weight: 214,22 g/mol Siedepunkt: 371 °C (1013 hPa) Schmelzpunkt: 170 °C |
MDL:
MFCD00002275 CAS-Nummer: 2051-85-6 EINECS: 218-131-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...