3-Chinolincarbaldehyd

Lieferant: Molekula

28695564-1G
MOLE28695564-1GEA 0 EUR
MOLE28695564-1G
3-Chinolincarbaldehyd
3-Chinolincarbaldehyd
Formel: C₁₀H₇NO
Lagertemperatur: Kühlschrank
MDL: MFCD00006768
CAS-Nummer: 13669-42-6
EINECS: 237-147-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR