4'-Hydroxy-3'-nitroacetophenon
Lieferant: Molekula
Synonyme:
1-(4-Hydroxy-3-nitrophenyl)-1-ethanone
28698508-5G
MOLE28698508-5GEA
0
EUR
MOLE28698508-5G
4'-Hydroxy-3'-nitroacetophenon
4'-Hydroxy-3'-nitroacetophenon
Formel:
C₈H₇NO₄ Molecular Weight: 181,15 g/mol |
MDL:
MFCD00017002 CAS-Nummer: 6322-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...