(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Lieferant: Molekula

Synonyme: (S)-(-)-BINAP

28868113-1G
MOLE28868113-1GEA 68.7 EUR
MOLE28868113-1G
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formel: C₄₄H₃₂P₂
Molecular Weight: 622,69 g/mol
Schmelzpunkt: 241…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010805
CAS-Nummer: 76189-56-5
Merck Index: 13,01223

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR