Acetylcholinchlorid
Lieferant: Molekula
Synonyme:
Acetylcholiniumchlorid, (2-Acetoxyethyl)trimethylammoniumchlorid
29010529-100G
MOLE29010529-100GEA
242
EUR
MOLE29010529-100G
Acetylcholinchlorid
Acetylcholinchlorid
Formel:
H₃CC(O)O(CH₂)₂N(CH₃)₃Cl Molecular Weight: 181,66 g/mol Schmelzpunkt: 151 °C Dichte: 1,263 g/cm³ (20 °C) |
MDL:
MFCD00011698 CAS-Nummer: 60-31-1 EINECS: 200-468-8 Merck Index: 13,00089 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...