Monomethyl-5-nitroisophthalat
Lieferant: Molekula
Synonyme:
5-Nitro-1,3-benzoldicarbonsäuremonomethylester, 5-Nitroisophthalsäuremonomethylester
29299560-100G
MOLE29299560-100GEA
0
EUR
MOLE29299560-100G
Monomethyl-5-nitroisophthalat
Monomethyl-5-nitroisophthalat
Formel:
C₉H₇NO₆ Molecular Weight: 225,16 g/mol Schmelzpunkt: 177…180 °C |
MDL:
MFCD00009793 CAS-Nummer: 1955-46-0 EINECS: 217-793-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...