N-Phenylmaleimid

Lieferant: Molekula

29392063-100G
MOLE29392063-100GEA 0 EUR
MOLE29392063-100G
N-Phenylmaleimid
N-Phenylmaleimid
Formel: C₁₀H₇NO₂
Siedepunkt: 299 °C (1013 hPa)
Schmelzpunkt: 84…86 °C
MDL: MFCD00005502
CAS-Nummer: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,07452

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR