Serotonin Hydrochlorid
Lieferant: Molekula
Synonyme:
5-Hydroxytryptamine Hydrochloride
29525281-1G
MOLE29525281-1GEA
71.6
EUR
MOLE29525281-1G
Serotonin Hydrochlorid
Serotonin Hydrochlorid
Formel:
C₁₀H₁₃ClN₂O Molecular Weight: 212,68 g/mol |
MDL:
MFCD00012686 CAS-Nummer: 153-98-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...