2-Phenoxyanilin

Lieferant: Molekula

Synonyme: 1-Amino-2-phenoxybenzol

29529332-25G
MOLE29529332-25GEA 61 EUR
MOLE29529332-25G
2-Phenoxyanilin
2-Phenoxyanilin
Formel: C₁₂H₁₁NO
Molecular Weight: 185,23 g/mol
Siedepunkt: 170 °C (18 mmHg)
Schmelzpunkt: 46…48 °C
MDL: MFCD00035765
CAS-Nummer: 2688-84-8
EINECS: 220-254-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR