2-Aminoimidazolhemisulfat

Lieferant: Molekula

29958183-25G
MOLE29958183-25GEA 0 EUR
MOLE29958183-25G
2-Aminoimidazolhemisulfat
2-Aminoimidazolhemisulfat
Formel: C₆H₁₂N₆O₄S
MDL: MFCD00013162
CAS-Nummer: 1450-93-7
EINECS: 215-918-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR