2-Oxazolidinon

Lieferant: Molekula

Synonyme: 2-Oxazolidone, Oxazolidin-2-one

29964445-100G
MOLE29964445-100GEA 210 EUR
MOLE29964445-100G
2-Oxazolidinon
2-Oxazolidinon
Formel: C₃H₅NO₂
Molecular Weight: 87,08 g/mol
MDL: MFCD00005268
CAS-Nummer: 497-25-6
EINECS: 207-840-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR