4-Aminobenzamidin Dihydrochlorid

Lieferant: Molekula

Synonyme: 4-Aminobenzoesäureamidin Dihydrochlorid, 4-Aminophenylamidin Dihydrochlorid

30099181-5G
MOLE30099181-5GEA 96.8 EUR
MOLE30099181-5G
4-Aminobenzamidin Dihydrochlorid
4-Aminobenzamidin Dihydrochlorid
Formel: C₇H₁₁Cl₂N₃
Molecular Weight: 208.09 g/mol
Schmelzpunkt: 290…291 °C
MDL: MFCD00013001
CAS-Nummer: 2498-50-2
EINECS: 219-692-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR