2-Methoxyphenethylamin

Lieferant: Molekula

30113132-25G
MOLE30113132-25GEA 0 EUR
MOLE30113132-25G
2-Methoxyphenethylamin
2-Methoxyphenethylamin
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
MDL: MFCD00008186
CAS-Nummer: 2045-79-6
EINECS: 218-066-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR