5-Chlor-2-nitrodiphenylamin
Lieferant: Molekula
Synonyme:
5-Chloro-2-nitro-N-phenylaniline
30181991-25G
MOLE30181991-25GEA
0
EUR
MOLE30181991-25G
5-Chlor-2-nitrodiphenylamin
5-Chlor-2-nitrodiphenylamin
Formel:
C₁₂H₉ClN₂O₂ Molecular Weight: 248,6700 g/mol |
MDL:
MFCD00007287 CAS-Nummer: 25781-92-4 EINECS: 247-261-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...