N-Methyl-N-nitroso-4-toluolsulfonamid
Lieferant: Molekula
30261311-25G
MOLE30261311-25GEA
210
EUR
MOLE30261311-25G
N-Methyl-N-nitroso-4-toluolsulfonamid
N-Methyl-N-nitroso-4-toluolsulfonamid
Formel:
C₈H₁₀N₂O₃S Molecular Weight: 214,24 g/mol |
MDL:
MFCD00002050 CAS-Nummer: 80-11-5 EINECS: 201-252-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...