D-Methionin

Lieferant: Molekula

Synonyme: (R)-(+)-2-Amino-4-(methylmercapto)buttersäure, (R)-(+)-Methionin, (R)-(+)-2-Amino-4-(methylthio)buttersäure, D(+)-Methionin, H-D-Met-OH

30263029-25G
MOLE30263029-25GEA 0 EUR
MOLE30263029-25G
D-Methionin
D-Methionin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.21 g/mol
Siedepunkt: 278
Schmelzpunkt: 136
MDL: MFCD00002622
CAS-Nummer: 348-67-4
EINECS: 206-483-6
Merck Index: 13,06004

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR