N-Benzylglycinethylester
Lieferant: Molekula
Synonyme:
Ethyl 2-(benzylamino)acetate
30356549-25G
MOLE30356549-25GEA
0
EUR
MOLE30356549-25G
N-Benzylglycinethylester
N-Benzylglycinethylester
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,25 g/mol |
MDL:
MFCD00009174 CAS-Nummer: 6436-90-4 EINECS: 229-218-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...