1-Acetylindol

Lieferant: Molekula

Synonyme: N-Acetylindol, 1-Indolylmethylketon, 1-Indolyl methyl ketone, 1-(1-Indolyl)-1-ethanon, Indol-1-ylmethylketon

30505578-1G
MOLE30505578-1GEA 0 EUR
MOLE30505578-1G
1-Acetylindol
1-Acetylindol
Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
Siedepunkt: 97,5…101 °C (0.1 mmHg)
Dichte: 1,387 g/cm³ (20 °C)
MDL: MFCD00038005
CAS-Nummer: 576-15-8
EINECS: 209-396-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR