α,α,α,α',α',α'-Hexafluor-2,5-xylidin
Lieferant: Molekula
Synonyme:
2,5-Bis(trifluoromethyl)aniline
30543599-1G
MOLE30543599-1GEA
0
EUR
MOLE30543599-1G
α,α,α,α',α',α'-Hexafluor-2,5-xylidin
α,α,α,α',α',α'-Hexafluor-2,5-xylidin
Formel:
C₈H₅F₆N Siedepunkt: 173…177 °C (1013 hPa) Dichte: 1,479 g/cm³ (20 °C) |
MDL:
MFCD00074940 CAS-Nummer: 328-93-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...