6-Chlorindol

Lieferant: Molekula

Synonyme: 6-Chloro-1H-indole

30799988-5G
MOLE30799988-5GEA 110 EUR
MOLE30799988-5G
6-Chlorindol
6-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151,6 g/mol
Schmelzpunkt: 90 °C
MDL: MFCD00005681
CAS-Nummer: 17422-33-2
EINECS: 241-449-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR