(2-Methoxyphenyl)acetonitril

Lieferant: Molekula

Synonyme: 2-Methoxybenzylcyanid

31013111-100G
MOLE31013111-100GEA 0 EUR
MOLE31013111-100G
(2-Methoxyphenyl)acetonitril
(2-Methoxyphenyl)acetonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00001902
CAS-Nummer: 7035-03-2
EINECS: 230-314-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR