D(+)-Fucose

Lieferant: Molekula

31642524-1G
MOLE31642524-1GEA 0 EUR
MOLE31642524-1G
D(+)-Fucose
D(+)-Fucose
Formel: C₆H₁₂O₅
MDL: MFCD00135603
CAS-Nummer: 3615-37-0
EINECS: 222-792-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR