Thymin
Lieferant: Molekula
Synonyme:
5-Methyluracil, 2,4-Dihydroxy-5-methylpyrimidin
31681796-25G
MOLE31681796-25GEA
0
EUR
MOLE31681796-25G
Thymin
Thymin
Formel:
C₅H₆N₂O₂ Molecular Weight: 126.11 g/mol Siedepunkt: 389 Schmelzpunkt: 218 Dichte: 1.454 g/cm³ (25 °C) |
MDL:
MFCD00006026 CAS-Nummer: 65-71-4 EINECS: 200-616-1 Merck Index: 12,09539 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...