β-Cyclodextrin

Lieferant: Molekula

Synonyme: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

31870153-100G
MOLE31870153-100GEA 209 EUR
MOLE31870153-100G
β-Cyclodextrin
β-Cyclodextrin
Formel: C₄₂H₇₀O₃₅
Molecular Weight: 1134,98 g/mol
MDL: MFCD00078139
CAS-Nummer: 7585-39-9
EINECS: 231-493-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR