2-Brom-5-fluoranilin

Lieferant: Molekula
Achtung

32061408
MOLE32061408EA 0 EUR
MOLE32061408
2-Brom-5-fluoranilin
2-Brom-5-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD00070750
CAS-Nummer: 1003-99-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR